Learn More
Medchemexpress LLC Diiodo(p-cymene)ruthenium(II) dimer | 90614-07-6 | MFCD08064027 | ≥98.0% | C20H28I4Ru2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY79986500MG
Encompass_Preferred
Diiodo(p-cymene)ruthenium(II) dimer is an organometallic compound commercially available for research and bulk quantities. This red-brown to dark brown powder has a molecular formula of C20H28I4Ru2 and a molecular weight of 978.19 g/mol. It exhibits a melting point of 234-236 °C, is insoluble in water, but soluble in chlorinated solvents like dichloromethane and chloroform. Optimal storage involves keeping it at 2-8°C under an inert atmosphere, protected from light and moisture.
  • Precursor for highly active catalysts in asymmetric transfer hydrogenation reactions.
  • Potential as an anticancer agent in medicinal chemistry and drug development.
  • Inhibits the PI3K/Akt/mTOR signaling pathway for anticancer effects.
  • Used in high-throughput screening workflows for catalyst development.
  • Presents as a red-brown to dark brown solid.
  • Characterization confirmed by 1H and 13C NMR spectroscopy.
Catalog No. 50-004-33187