Learn More
Medchemexpress LLC Dilauryl thiodipropi 500g
SDP
Supplier:  Medchemexpress LLC HYW099538500G
Encompass_Preferred
Dilauryl thiodipropionate (DLTDP) which is a sulfur-containing antioxidant commonly used to stabilize polymers and plastics against degradation caused by heat oxygen and UV light acts to scavenge free radicals and others that may cause polymer chain scission and cross-linked active substances in addition DLTDP has been used as an additive to lubricants oils and other industrial fluids to improve their oxidation stability the long hydrocarbon chain in DLTDP makes it low volatility and compatible with many materials and Good compatibility with substrates
Catalog No. 50-002-05192