Learn More
Medchemexpress LLC Dilazep dihydrochloride | 35898-87-4 | 604.69 g/mol | C31H44N2O10 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10095710MM 1ML
Encompass_Preferred
Dilazep dihydrochloride is a research reagent that inhibits adenosine uptake and exhibits cerebral and coronary vasodilatory activity. It is supplied for laboratory research use and is identified by CAS 35898-87-4, molecular formula C31H44N2O10, and molecular weight 604.69 g/mol.
  • Inhibits adenosine uptake for pharmacological studies.
  • Demonstrates cerebral and coronary vasodilation effects.
  • Chemical identifiers included for accurate compound tracking.
  • Suitable for in vitro and biochemical assays when used per protocols.
  • Stored under recommended conditions to maintain stability.
Catalog No. 50-002-12269