Learn More
Selleck Chemical LLC Diltiazem E7160-100MG
SDP
Supplier:  Selleck Chemical LLC E7160100MG
Encompass_Preferred
Diltiazem (Dilzen CRD-401) is a benzothiazepine an antagonist of Ca2 channel inhibiting L-type Ca currents via CaV1 2 channels in cardiac and vascular smooth muscle It can be used in research of cardiac arrhythmia hypertension and angina pectoris
Catalog No. 50-000-42207