Learn More
Medchemexpress LLC Diltiazem | 42399-41-7 | 99.7% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0632500MG
Encompass_Preferred
Diltiazem is an orally active L-type Ca2+ channel blocker. It exhibits antihypertensive and antiarrhythmic effects and is utilized in research for cardiac arrhythmia, hypertension, and angina pectoris.
  • Acts as an L-type calcium channel blocker
  • Shows antihypertensive and antiarrhythmic effects
  • Useful for cardiac arrhythmia research
  • Applicable for hypertension and angina pectoris research
  • Demonstrated antiproliferative activity in certain cell lines
  • Inhibits P-glycoprotein-mediated multidrug resistance
  • Prevents aortic aneurysm formation in animal models
Catalog No. 50-004-05936