Learn More
Selleck Chemical LLC Diltiazem HCl S1865-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S186510MM/1ML
Encompass_Preferred
Diltiazem (Tiazac RG 83606) HCl is a benzothiazepine derivative with vasodilating action due to its antagonism of the actions of the calcium ion in membrane functions
Catalog No. 50-310-9288