Learn More
Medchemexpress LLC Diludine (Et-HE) | 1149-23-1 | MFCD00005951 | 99.7% | 253.29 g/mol | C13H19NO4 | 1 KG

Supplier: Medchemexpress LLC HYW0179601KG
Diludine (Et-HE) is a 1,4-dihydropyridine derivative (Hantzsch ester) used as an antioxidant in biological studies and as a hydride donor in organic synthesis. It is supplied as an off-white to yellow solid with documented solubility and storage recommendations for research use.
- High purity (99.71%).
- Off-white to yellow solid form.
- Soluble in DMSO at 50 mg/mL; ultrasonic assistance recommended.
- Stable at room temperature; protect from light; stock solutions: -80°C (2 years), -20°C (1 year).
- Used as an antioxidant in animal studies and as a hydride source in organic reactions.
- CAS number 1149-23-1 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.