Learn More
Cayman Chemical DIM-C-pPhCO2Me 10mg
SDP
Supplier:  Cayman Chemical 3754010 MG
Encompass_Preferred
An NR4A1 antagonist; decreases the expression of the mRNA encoding integrin β1 in MIA PaCa-2, PANC-1, RKO, and SW480 cells at 15 and 20 µM; decreases the migration and adhesion of MIA PaCa-2 and SW480 cells at 15 and 20 µM
Catalog No. 50-291-5542