Learn More
Selleck Chemical LLC DIM-C-pPhOH-5mg
SDP
Supplier:  Selleck Chemical LLC S67995MG
Encompass_Preferred
DIM-C-pPhOH (C-DIM8, CDIM8) is a nuclear receptor 4A1 (NR4A1)(Nur77) antagonist. DIM-C-pPhOH inhibits tumor growth and mTOR signaling, induces apoptosis and cellular stress. DIM-C-pPhOH reduces cell proliferation with IC50 of 13.6 μM and 13.0 μM for ACHN cells and 786-O cells, respectively.
Catalog No. 50-238-9879