Learn More
Medchemexpress LLC Dimaprit dihydrochloride | 23256-33-9 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB147810MM/1ML
Encompass_Preferred
Dimaprit dihydrochloride is a selective histamine H2 receptor agonist that also inhibits nNOS with an an IC50 of 49 μM. It has the ability to stimulate gastric acid secretion. This compound shows less than 0.0001% the activity of histamine on H1-receptors.
  • Selective histamine H2 receptor agonist.
  • Inhibits nNOS with an IC50 of 49 μM.
  • Can stimulate gastric acid secretion.
  • Less than 0.0001% activity on H1-receptors compared to histamine.
Catalog No. 50-004-11140