Learn More
Medchemexpress LLC Dimaprit dihydrochloride | 23256-33-9 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1478
Encompass_Preferred
Dimaprit dihydrochloride is a selective histamine H2 receptor agonist, and it also inhibits nNOS with an IC50 of 49 μM. This product can stimulate gastric acid secretion.
  • Selective histamine H2 receptor agonist.
  • Inhibits nNOS with an IC50 of 49 μM.
  • Stimulates gastric acid secretion.
  • Has less than 0.0001% the activity of histamine on H1-receptors.
  • Causes dose-dependent falls in blood pressure in cats.
  • Causes vasodilatation in the femoral vascular bed.
Catalog No. 50-004-14213