Learn More
Medchemexpress LLC Dimboa | 15893-52-4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN743210MG
Encompass_Preferred
DIMBOA, a benzoxazinoid, serves as a chemical defense in graminaceous plants like maize, wheat, and rye. It inhibits the growth of bacteria such as *Staphylococcus aureus* and *Escherichia coli*, and fungi including *Saccharomyces cerevisiae*. Demonstrating antioxidant activity, DIMBOA shows potent free-radical scavenging and inhibits the biosynthesis of toxic trichothecenes, thereby reducing plant susceptibility to scab. It also exhibits cytotoxicity to plant cells.
  • Functions as a chemical defense in graminaceous plants
  • Inhibits growth of bacteria and fungi
  • Potent free-radical scavenging and antioxidant properties
  • Inhibits biosynthesis of toxic trichothecenes
  • Reduces plant susceptibility to scab
  • Exhibits cytotoxicity to plant cells
  • For research use only
Catalog No. 50-004-19574