Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimedone (5,5-dimethylcyclohexane-1,3-dione) | 126-81-8 | MFCD00001588 | 99.7% | 140.18 g/mol | C8H12O2 | 1000 G
SDP

Supplier:  Medchemexpress LLC HYW0157401000G

Encompass_Preferred

Dimedone (5,5-dimethylcyclohexane-1,3-dione) is a small-molecule biochemical reagent used in laboratory research. It is a cyclic diketone (CAS 126-81-8) with formula C8H12O2 and molecular weight 140.18 g/mol. The material is supplied for research use only and is provided at high stated purity.

  • High purity (99.7%).
  • CAS number 126-81-8.
  • Chemical formula C8H12O2.
  • Molecular weight 140.18 g/mol.
  • Soluble in DMSO.
  • Available in multiple bulk pack sizes for laboratory use.

Catalog No. 50-003-00676


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.