Learn More
Selleck Chemical LLC Dimercaprol S4446-1g
SDP
Supplier:  Selleck Chemical LLC S44461G
Encompass_Preferred
Dimercaprol (Bal 2 3-Dimercapto-1-propanol Dithioglycerol) is an acrolein scavenger that inhibits HIV-1 tat activity viral production and infectivity in vitro
Catalog No. 50-311-0247