Learn More
Medchemexpress LLC Dimercaprol | 59-52-9 | 98.0% | 1 G

Supplier: Medchemexpress LLC HYB12851G
Dimercaprol (2,3-Dimercapto-1-propanol) is an anti-heavy metal-poisoning agent that also exhibits anti-HIV activity. It is used for research on poisoning by arsenic, mercury, gold, lead, antimony, and other toxic metals.
- Anti-heavy metal-poisoning agent
- Exhibits anti-HIV activity
- Can be used for the study of arsenic, mercury, gold, lead, antimony, and other toxic metal poisoning
- Can significantly protect PC-12 cells from Acrolein-mediated cell death in a dose-dependent manner
- Decreases mercury contents in liver and kidney in Wistar rats exposed to HgCl2
- Appearance: Liquid (Density: 1.239 g/cm³)
- Color: Colorless to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.