Learn More
Medchemexpress LLC Dimethoate-d6 | 1219794-81-6 | 99.86% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1946S1MG
Encompass_Preferred
Dimethoate-d6 is the deuterium-labeled version of Dimethoate. Dimethoate is an organophosphate insecticide and acaricide, acting as an orally active acetylcholinesterase inhibitor. It is known to induce reactive oxygen species (ROS), DNA damage, and cell apoptosis in vivo, and has been observed to affect the immune system in mice.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11250