Learn More
Medchemexpress LLC Dimethyl adipate-d8 | 52089-64-2 | MFCD12022192 | 98.0% | 182.24 g/mol | C8H6D8O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW015301S25MG
Encompass_Preferred
Dimethyl adipate-d8 is the deuterium-labeled analogue of dimethyl adipate, intended as an isotope-labeled analytical standard for use in mass spectrometry and other quantitative analytical methods. It is supplied as a high-purity, research-grade reagent for laboratory use only and is not for human or clinical applications.
  • Suitable as an internal standard for GC-MS and LC-MS quantitation.
  • Deuterium labeling (d8) provides a clear mass shift for accurate analyte measurement.
  • High purity (98.0%) minimizes background and matrix interferences.
  • Offered in multiple small sizes to support analytical workflows.
  • Accompanied by datasheet and certificate of analysis for quality verification.
Catalog No. 50-003-03670