Learn More
Medchemexpress LLC Dimethyl adipate-d8 | 52089-64-2 | MFCD12022192 | 98.0% | 182.24 g·mol⁻¹ | C8H6D8O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW015301S250MG
Encompass_Preferred
Dimethyl adipate-d8 is the deuterium-labeled form of dimethyl adipate (hexanedioic acid dimethyl ester) intended as an analytical reference standard for research use. It is used as an isotopically labeled tracer and internal standard to support accurate quantitation and metabolic tracing in chemical and biological analyses.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • High purity (98.0%) and isotopic enrichment for reliable results.
  • Useful as a tracer in metabolic and labeling studies.
  • Provided as a stable reagent for analytical applications.
Catalog No. 50-003-02930