Learn More
Medchemexpress LLC Dimethyl dodecanedioate | 1731-79-9 | 99.46% | 258.36 | 500 G

Supplier: Medchemexpress LLC HYW013359500G
Dimethyl dodecanedioate is an ester product.
- Available in sizes of 100 g and 500 g, both in-stock.
- Can be ordered for bulk inquiry.
- Colorless to off-white appearance.
- Solid below 27°C, liquid above 32°C.
- Shipping at room temperature in continental US.
- Storage: 3 years at room temperature; in solvent, 2 years at -80°C and 1 year at -20°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.