Learn More
Medchemexpress LLC Dimethyl phthalate (Ring-d4) | 93951-89-4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN7106S5MG
Encompass_Preferred
Dimethyl phthalate (Ring-d4) is a deuterium-labeled form of Dimethyl phthalate, which is known as an endocrine disruptor and a ubiquitous pollutant from the phthalate esters (PAEs) group. It is commonly used as a plasticizer to make rigid polyvinylchloride (PVC) resins more flexible.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound.
  • Suitable for research use only.
Catalog No. 50-004-19663