Learn More
Medchemexpress LLC Polydimethylsiloxane | 63148-62-9 | MFCD00084411 | >6,800 Da | [Si(CH3)2O]n | 25 ML
SDP
Supplier:  Medchemexpress LLC HY104035E25ML
Encompass_Preferred
Dimethyl silicone oil (polydimethylsiloxane) is a clear, colorless silicone fluid with a kinematic viscosity of 100 cSt at 25 °C (CAS 63148-62-9). It is an inert, low-volatility laboratory reagent used where a defined silicone medium is required, including as viscosity standards, lubricants, anti-foaming agents, and hydrophobic treatments. Supplied in a 25 mL bottle for small-scale laboratory use.
  • Viscosity 100 cSt at 25 °C
  • Clear, colorless liquid
  • Low volatility and chemically inert
  • Suitable as a viscosity standard and general-purpose silicone fluid
  • Useful as a lubricant, anti-foaming agent, or hydrophobic barrier
  • Packaged for small-scale laboratory use (25 mL)
Catalog No. 50-002-43356