Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl sulfate, 77-78-1, MFCD00008416, 100 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 41610100ML
Encompass
Linear Formula: (CH3O)2SO2, Molecular Weight: 126.13, puriss. p.a., ≥99.0% (GC), Synonym: Sulfuric acid dimethyl ester.
Catalog No. NC1610601