Learn More
Supply Solutions Dimethyl sulfoxide, 67-68-5, MFCD00002089, 100mL
Supplier:  Supply Solutions SIGMA D8418100ML
Encompass_Preferred
Linear Formula (CH3)2SO, Molecular Weight 78.13, bp 189 C (lit.), mp 16-19 C (lit.), for molecular biology, Synonym: DMSO
Catalog No. 50-161-3339
 Restricted Item