Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl sulfoxide Hybri-Max, 99.7, 67-68-5, MFCD00002089, 5x10mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D26505X10ML
Encompass
Linear Formula (CH3)2SO, ≥99.7%, Molecular Weight 78.13, Hybri-Max™, sterile-filtered, BioReagent, Synonym: DMSO
Catalog No. 50-165-7122
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.