Learn More
Medchemexpress LLC Dimethyl sulfoxide | 67-68-5 | MFCD00002089 | 100.0% | 78.13 g/mol | C2H6OS | 50mL
SDP
Supplier:  Medchemexpress LLC HYY0320GL50ML
Encompass_Preferred
Dimethyl sulfoxide (DMSO) (GMP Like) is the GMP Like class Dimethyl sulfoxide (HY-Y0320C) Dimethyl sulfoxide is an aprotic solvent that can dissolve water-insoluble therapeutic and toxic agents Dimethyl sulfoxide has a strong affinity for water and has the ability to rapidly penetrate or enhance the penetration of other substances through biological membranes Dimethyl sulfoxide also has potential free radical scavenging and anticholinesterase effects and may affect coagulation activity Dimethyl sulfoxide also induces histamine release from mast cells but is thought to have low systemic toxicity[1]
Catalog No. 50-002-68200