Learn More
Medchemexpress LLC Dimethyl terephthalate | 120-61-6 | MFCD00008440 | 99.9% | 194.18 g/mol | C10H10O4 | 1kg
SDP
Supplier:  Medchemexpress LLC HYY03831KG
Encompass_Preferred
Dimethyl terephthalate is a low toxicity low volatility industrial chemical widely used in the production of polyester fibers and films[1][2]
Catalog No. 50-002-69210