Learn More
Medchemexpress LLC Dimethylamino-PEG3 100mg
SDP
Supplier:  Medchemexpress LLC HYW096150100MG
Encompass_Preferred
Dimethylamino-PEG3 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05662