Learn More
Medchemexpress LLC Dimethylcurcumin | 52328-98-0 | 98.0% | 396.43 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY15194
Encompass_Preferred
Dimethylcurcumin (ASC-J9) is an androgen receptor degradation enhancer that effectively suppresses castration-resistant prostate cancer cell proliferation and invasion. This product is an analytical standard intended for research and analytical applications, not for patient use.
  • Enhances androgen receptor degradation
  • Suppresses castration-resistant prostate cancer cell proliferation
  • Suppresses castration-resistant prostate cancer cell invasion
  • Intended for research and analytical applications
Catalog No. 50-003-64506