Learn More
Medchemexpress LLC Dimethylcurcumin | 52328-98-0 | C23H24O6 | 100 MG

Supplier: Medchemexpress LLC HY15194100MG
Dimethylcurcumin (ASC-J9) is an androgen receptor degradation enhancer that effectively suppresses castration-resistant prostate cancer cell proliferation and invasion. It works by selectively promoting AR degradation by disrupting the interaction between AR and AR coregulators, and it reduces the aggregation of AR-112Q in cells.
- Enhances androgen receptor degradation
- Suppresses castration-resistant prostate cancer cell proliferation and invasion
- Promotes AR degradation by disrupting interaction between AR and AR coregulators
- Reduces aggregation of AR-112Q in cells
- High purity (98.01%)
- Molecular weight of 396.43
- Solid appearance
- Yellow to orange color
- Soluble in DMSO (≥ 50 mg/mL)
- Suitable for in vivo preparation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.