Learn More
Medchemexpress LLC Dimethylcurcumin | 52328-98-0 | C23H24O6 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15194
Encompass_Preferred
Dimethylcurcumin is an analytical standard intended for research and analytical applications. It acts as an androgen receptor degradation enhancer, effectively suppressing castration-resistant prostate cancer cell proliferation and invasion.
  • Analytical standard for research
  • Used in analytical applications
  • Androgen receptor degradation enhancer
  • Suppresses castration-resistant prostate cancer cell proliferation
  • Suppresses castration-resistant prostate cancer cell invasion
Catalog No. 50-003-74534