Learn More
Medchemexpress LLC Dimethylcurcumin (ASC-J9) | 52328-98-0 | MFCD12912341 | 98.0% | 396.43 | C23H24O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1519410MG
Encompass_Preferred
Dimethylcurcumin (ASC-J9) is a small-molecule androgen receptor (AR) degradation enhancer used in research to suppress castration-resistant prostate cancer cell proliferation and invasion. It selectively promotes degradation of AR and AR splice variants by disrupting AR-coregulator interactions, leading to reduced AR-driven gene expression. Supplied as a powder for laboratory use with specified solvent solubility and storage conditions.
Catalog No. 50-002-87512