Learn More
Medchemexpress LLC Dimethyldioctadecylammonium (bromide) | 3700-67-2 | ≥99.0% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY131119
Encompass_Preferred
Dimethyldioctadecylammonium bromide is a synthetic cationic lipid primarily utilized in gene delivery and vaccine development. It is characterized by a positively charged ammonium head group and two long hydrophobic tails. These structural features enable it to form liposomes and other lipid-based nanoparticles, which are effective for delivering genetic material into cells. Beyond its biotechnological applications, it also finds use in surfactants, emulsifiers, and fabric softeners. Due to its potential for toxicity and irritation, caution is advised during handling. This product is intended for research use only and not for sale to patients.
  • Used in gene delivery and vaccine development
  • Forms liposomes and other lipid-based nanoparticles
  • Effective for delivering genetic material into cells
  • Used as surfactants, emulsifiers, and fabric softeners
  • Intended for research use only
Catalog No. 50-003-42077