Learn More
Strem, An Ascensus Company CAS# 756-79-6. 500g. Dimethylmethylphosphonate, 97%. MFCD00008349
SDP
Supplier:  Strem, An Ascensus Company 931523500G
Encompass
CAS# 756-79-6. 500g. Dimethylmethylphosphonate, 97%. MFCD00008349. Molecular Weight: 124.08. Molecular Formula: (CH3O)2P(O)(CH3). Color/form: colorless liq. Strem# 93-1523. http://www.strem.com/catalog/v/93-1523/
Catalog No. 50-901-14206