Learn More
Abcam Dimethylo x aloylglycine, Prolyl hydro x ylase (PHD) inhibitor, 50MG
Supplier:  Abcam AB14158650MG
Encompass_Preferred
MW 250.26 Da, Purity >98%. Cell-permeable prolyl hydroxylase (PHD) inhibitor. Upregulates HIF activity. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6508