Learn More
Selleck Chemical LLC dimyristoyl lecithin E2870-5MG
SDP
Supplier:  Selleck Chemical LLC E28705MG
Encompass_Preferred
dimyristoyl lecithin (DMPC) is a synthetic phospholipid used in liposomes 1 2-Dimyristoyl-sn-glycero-3-phosphocholine is used for the study of lipid monolayers and bilayers
Catalog No. 50-000-39734