Learn More
Medchemexpress LLC Dinaphtho[2,1-d:1',2'-f]dioxaphosphepin, 4-hydroxy-, 4-oxide, (11bR)- | 39648-67-4 | 99.9% | 348.29 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYI0718
Encompass_Preferred
(R)-1,1'-Binaphthyl-2,2'-diyl hydrogenphosphate is a biochemical reagent suitable for life science research. It functions as a chiral ligand in various organic reactions.
  • Used in hydrocarboxylation reactions.
  • Forms complexes with rhodium to mediate asymmetric dipolar cyclodition of diazo compounds.
  • Resolves racial amines that are difficult to separate.
  • Palladium and rhodium derivatives utilized in asymmetric hydrocarboxylations and dipolar cycloditions.
  • Employed in chiral Bronsted acid catalyzed enantioselective Mannich reactions.
Catalog No. 50-004-31344