Learn More
Dino Lite USB CapillaryScope AF4515-N4UT - 1.3MP, 400x - 470x Optical Magnification, WF-20 Compatible
Supplier:  Dino Lite AF4515N4UT
Encompass_Preferred
The AF4515-N4UT Dino-Lite CapillaryScope uses a magnification range of 400x - 470x. It is intended for inspection of microcirculation (microcapillary function) of the blood vessels under fingernails. The curved cap (N3C-N) provides easy focusing and stability over the fingernail viewing area. The sealed cap (N3C-C) can protect the inner component and lens of the Dino-Lite and offer options for viewing skin with oil immersion. The MicroTouch feature is a touch sensitive area which can be used to capture images instead of using a keyboard and mouse. The built-in adjustable polarizer provides additional convenience as well as imaging ability by eliminating glare and viewing through polarized light. The new modular wireless-ready design allows for USB connection or a quick swap to wireless with a WF-20 adapter (sold separately) making it a completely cordless and portable microscope.
Catalog No. NC1914728