Learn More
Dino Lite VGA Digital Microscope AM5216ZTL- 720p, 5x - 140x Optical Magnification, Polarized Light, Long Working Distance
Supplier:  Dino Lite AM5216ZTL
Encompass_Preferred
The Dino-Lite AM5216ZTL Edge Series features a High Definition VGA connection and improved optics for sharper images. It easily allows for the observation of objects at a magnification range of 5x - 140x. Capable of 60 frames per second at 720p (1280 x 720), this model is ideal for observing specimens effectively with minimum lag while in motion. No computer or software required for this model that connects directly to a monitor or TV with VGA connections. Polarization can be switched on/off or can be adjusted to offer full or half polarization for controlling glare.
Catalog No. NC1573622