Learn More
Dino Lite RK-06A Multi-purpose Tabletop Stand
Supplier:  Dino Lite RK06A
Encompass_Preferred
The Dino-Lite RK-06A tabletop stand is designed for general purpose and built with quality. With smooth focus adjustment and quick vertical movement, the RK-06A eases the task of focusing when using Dino-Lite microscopes. The ease-of-assembly and adaptable extension arms (2) design add further mobility and flexibility to user’s applications. For users that require additional working distance, the optional RK-06-PL extension pole is sold separately.
Catalog No. NC1636213