Learn More
Electron Microscopy Sciences This Dino-Lite Edge, Superior images at 10x - 140x magnification
SDP
Supplier:  Electron Microscopy Sciences AF7115MZTL
Encompass_Preferred
This Dino-Lite Edge is a Longer Working Distance (LWD) version of Dino-Lite 5 MP Edge series, delivering superior images at 10x - 140x magnification, with the best illumination flexibility. It was designed with plenty of space under which to work. It uses fluent and crisp live imaging through the use of cutting-edge optics, advanced 5 MP sensor, and low-loss MJPEG compression to provide long working distance. For better adaptability, the innovative Flexible LED Control (FLC) enables partial illumination to enhance edge contrast with intensity adjustability. With its rugged metal housing, built-on adjustable polarizer, and interchangeable front caps, this model is the tool of choice for a wide range of applications.
Catalog No. NC2825148