Learn More
Selleck Chemical LLC Dinoprost tromethamine S5056-10mg
SDP
Supplier:  Selleck Chemical LLC S505610MG
Encompass_Preferred
Dinoprost Tromethamine (Dinolytic PGF2-alpha tham Zinoprost Prostin F2 alpha Dinoprost Trometamol) is a synthetic analogue of the naturally occurring prostaglandin F2 alpha which stimulates myometrial activity relaxes the cervix inhibits corpus luteal steroidogenesis and induces luteolysis by direct action on the corpus luteum
Catalog No. 50-311-2050