Learn More
Medchemexpress LLC Dinoprost (tromethamine salt) | 38562-01-5 | 98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12956A50MG
Encompass_Preferred
Dinoprost tromethamine salt is an orally active, potent prostaglandin F (PGF) receptor (FP receptor) agonist. It is a luteolytic hormone produced locally in the endometrial luminal epithelium and corpus luteum (CL), playing a key role in the onset and progression of labor.
  • Orally active
  • Potent prostaglandin F (PGF) receptor (FP receptor) agonist
  • Luteolytic hormone produced locally in the endometrial luminal epithelium and corpus luteum (CL)
  • Plays a key role in the onset and progression of labor
  • Induces ER stress, autophagy, and apoptosis in goat luteal cells
  • Significantly increases the expression of GRP78 and UPR sensors
Catalog No. 50-003-39449