Learn More
TARGETMOL CHEMICALS INC DIO 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4041850MG
Encompass_Preferred
Also available in 5 mg 10 mg and bulk. Please contact Fisher for quotes. DiO (DiOC18(3)) is a fluorescent lipophilic tracer and can be widely used to label cells organelles liposomes viruses and lipoproteins. purity: 99%
Catalog No. 50-257-9991