Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dioctyl phthalate-d4 | 93952-13-7 | 99.7% | 394.58 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYW013755S5MG

Encompass_Preferred

Dioctyl phthalate-d4 is the deuterium labeled Dioctyl phthalate. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.

  • Liquid appearance (density: 0.991 g/cm³)
  • Colorless to light yellow
  • Formula: C24H34D4O4
  • Shipping at room temperature in continental US
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-54216


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.