Learn More
YMC America, Inc Diol 300A 3um 150x0.3mm 1/16
SDP
Supplier:  YMC America, Inc DL30S0315H0AU
Encompass
YMC-Pack Diol is a size exclusion chromatography column packed with dihydroxypropyl-bonded silica and available in four different pore sizes It exhibits high mechanical strength and is useful for molecular weight determination of proteins and sugars The 2 m and 3 m particle sizes provide high resolution and fast analysis
Catalog No. 50-008-80953