Learn More
YMC America, Inc Diol col 120A 5um 150x4.6mm
SDP
Supplier:  YMC America, Inc DL12S051546WT
Encompass_Preferred
YMC-Pack Diol is a size exclusion chromatography column packed with dihydroxypropyl-bonded silica and available in four different pore sizes It exhibits high mechanical strength and is useful for molecular weight determination of proteins and sugars The 2 m and 3 m particle sizes provide high resolution and fast analysis
Catalog No. 50-008-76957