Learn More
YMC America, Inc Diol-NP col 120A 5um 250x2mm
SDP
Supplier:  YMC America, Inc DN12S052502WT
Encompass_Preferred
YMC-Pack Diol-NP is a versatile alternative to silica for normal phase separations and provides much better reproducibility when compared to bare silica material It shows typical normal phase retention behavior when used with low-polarity mobile phases due to hydroxyl groups on the surface of the stationary phase acting as polar groups YMC-Pack Diol-NP can also be used in HILIC-mode with organic/water mobile phases
Catalog No. 50-008-80081