Learn More
Medchemexpress LLC Dioleyldimethylammonium chloride | 7212-69-3 | 95.9% | 582.47 g·mol⁻¹ | C38H76ClN | 100 MG

Supplier: Medchemexpress LLC HYW590549100MG
Dioleyldimethylammonium chloride (DODAC) is a cationic quaternary ammonium lipid used in research for liposome formulation and as a component in transfection reagents. Supplied with documented purity and storage recommendations, it is intended for laboratory research use only.
- Cationic lipid for liposome formulation and transfection applications.
- Purity 95.9% by supplied analysis.
- Chemical formula C38H76ClN; molecular weight 582.47 g·mol⁻¹.
- Available in small-scale quantities suitable for research use.
- Storage recommendations include sealed, dry storage at 4°C and low-temperature solvent storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.