Learn More
Medchemexpress LLC Dioleyldimethylammonium chloride | 7212-69-3 | 95.9% | 582.47 g/mol | C38H76ClN | 5 MG

Supplier: Medchemexpress LLC HYW5905495MG
Dioleyldimethylammonium chloride (DODAC) is a cationic lipid for research use that facilitates delivery of nucleic acids by forming lipid assemblies such as liposomes. It is provided as a characterized reagent suitable for laboratory transfection experiments.
- Cationic lipid suitable for nucleic acid delivery.
- Facilitates liposome formation and membrane interaction.
- Characterized by HNMR, RP-HPLC, and LCMS analytical methods.
- Purity approximately 95.9%.
- Molecular weight 582.47 g/mol and CAS number 7212-69-3.
- Available in small milligram quantities for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.