Learn More
Apexbio Technology LLC Dioscin 19057-60-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC N17235.1
Encompass_Preferred
Dioscin (CAS 19057-60-4) is a steroidal saponin isolated primarily from Dioscorea nipponica Makino Structurally it belongs to glycosidic steroid derivatives known for modulating various biological pathways Dioscin exerts bioactivity predominantly through regulation of cellular signaling cascades such as apoptosis induction anti-inflammatory responses and oxidative stress reduction Studies suggest that dioscin interacts with signaling pathways including PI3K/Akt ERK/MAPK and NF- B Due to these activities dioscin is commonly investigated for its potential anti-inflammatory anticancer and antioxidant effects making it relevant for biomedical research focused on chronic inflammation and cancer therapeutics
Catalog No. 50-000-63286